The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130046.1 | NP_001123518.1 | 3 - 4 | 330 | 96 |
NM_004591.2 | NP_004582.1 | 3 - 4 | 333 | 96 | |
GenBank mRNA | BC020698.1 | - | 3 - 4 | 330 | 96 |
D86955.1 | - | 3 - 4 | 321 | 96 | |
U64197.1 | - | 3 - 4 | 302 | 96 | |
U77035.1 | - | 3 - 4 | 263 | 96 |