The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271681.1 | NP_001258610.1 | 2 - 3 | 371 | 74 |
NM_013328.3 | NP_037460.2 | 2 - 3 | 371 | 74 | |
GenBank mRNA | AF087859.1 | - | 2 - 3 | 289 | 74 |
AF151351.1 | - | 2 - 3 | 169 | 74 | |
AK291913.1 | - | 2 - 3 | 206 | 74 | |
BC014868.1 | - | 2 - 3 | 262 | 74 | |
BC020553.1 | - | 2 - 3 | 296 | 74 | |
BU538765.1 | - | 2 - 3 | 246 | 74 | |
HY044355.1 | - | 2 - 3 | 372 | 74 | |
KU178615.1 | - | 2 - 3 | 141 | 74 |