Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPHK2
Entrez Gene ID
56848
Gene Name
sphingosine kinase 2
Gene Aliases
SK 2, SK-2, SPK 2, SPK-2
Chromosome Location
Chr.19: 48619291 - 48630407 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204158.2 NP_001191087.1 6 - 7 1063 72
NM_001204159.2 NP_001191088.1 6 - 7 1144 72
NM_001204160.2 NP_001191089.1 5 - 6 960 72
NM_001243876.1 NP_001230805.1 5 - 6 768 72
NM_020126.4 NP_064511.2 6 - 7 1240 72
XM_006723292.1 XP_006723355.1 5 - 6 930 72
XM_011527133.1 XP_011525435.1 6 - 7 1075 72
XM_011527134.1 XP_011525436.1 5 - 6 932 72
XM_017027008.1 XP_016882497.1 4 - 5 1646 72
XM_017027009.1 XP_016882498.1 5 - 6 651 72
GenBank mRNA AB100373.1 - 4 - 5 402 72
AF245447.1 - 4 - 5 772 72
AK074601.1 - - 1713 72
AK122735.1 - 7 - 8 944 72
AK300541.1 - 6 - 7 902 72
AL136701.1 - 6 - 7 1143 72
BC006161.1 - 6 - 7 1240 72
BC010671.1 - 5 - 6 960 72
EF107108.1 - 4 - 5 1768 72

View Full Product Details