Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UTP6
Entrez Gene ID
55813
Gene Name
UTP6, small subunit processome component
Gene Aliases
C17orf40, HCA66
Chromosome Location
Chr.17: 31862379 - 31901710 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018428.2 NP_060898.2 13 - 14 1209 100
XM_005258012.2 XP_005258069.1 12 - 13 1061 100
XM_011524997.2 XP_011523299.1 13 - 14 1180 100
XM_017024845.1 XP_016880334.1 13 - 14 1180 100
GenBank mRNA AF244135.1 - 13 - 14 1168 100
AF334386.1 - 13 - 14 1222 100
AK057202.1 - 13 - 14 1156 100
AK126564.1 - 6 - 7 1490 100
AK225448.1 - 13 - 14 1165 100
BC015454.1 - 13 - 14 1114 100
BC035325.1 - 13 - 14 1160 100

View Full Product Details