The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007792.1 | NP_001007793.1 | 5 - 6 | 419 | 66 |
NM_001012331.1 | NP_001012331.1 | 4 - 5 | 485 | 66 | |
NM_002529.3 | NP_002520.2 | 4 - 5 | 485 | 66 | |
GenBank mRNA | AK126428.1 | - | 5 - 6 | 490 | 66 |
AK290759.1 | - | 4 - 5 | 470 | 66 | |
AK312704.1 | - | 4 - 5 | 470 | 66 | |
BC062580.1 | - | 4 - 5 | 428 | 66 | |
BC136554.1 | - | 4 - 5 | 529 | 66 | |
BC144239.1 | - | 4 - 5 | 529 | 66 | |
CR936794.1 | - | 6 - 7 | 487 | 66 | |
KJ534902.1 | - | 4 - 5 | 429 | 66 | |
M23102.1 | - | 4 - 5 | 549 | 66 |