The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000816.3 | NP_000807.2 | 2 - 3 | 621 | 102 |
NM_198903.2 | NP_944493.2 | 2 - 3 | 621 | 102 | |
NM_198904.2 | NP_944494.1 | 2 - 3 | 621 | 102 | |
GenBank mRNA | AK299640.1 | - | 3 - 4 | 346 | 102 |
AK315200.1 | - | 2 - 3 | 273 | 102 | |
BC059389.1 | - | 2 - 3 | 442 | 102 | |
BC069348.1 | - | 2 - 3 | 357 | 102 | |
BC074795.2 | - | 2 - 3 | 295 | 102 | |
CD013896.1 | - | 2 - 3 | 399 | 102 | |
CD013897.1 | - | 2 - 3 | 393 | 102 | |
X15376.1 | - | 2 - 3 | 489 | 102 |