Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYOC
Entrez Gene ID
4653
Gene Name
myocilin
Gene Aliases
GLC1A, GPOA, JOAG, JOAG1, TIGR
Chromosome Location
Chr.1: 171635417 - 171652633 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000261.1 NP_000252.1 1 - 2 627 90
GenBank mRNA AF001620.1 - 1 - 2 627 90
AK097310.1 - 2 - 3 575 90
AK300924.1 - 2 - 3 503 90
AK300943.1 - 2 - 3 628 90
AK301297.1 - 2 - 3 548 90
AK315443.1 - 1 - 2 677 90
BC029261.1 - 1 - 2 665 90
D88214.1 - 1 - 2 669 90
FJ227302.1 - 1 - 2 627 90
FJ237046.1 - 1 - 2 627 90
FJ237047.1 - 1 - 2 627 90
KU178210.1 - 1 - 2 605 90
U85257.1 - 1 - 2 641 90

View Full Product Details