The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204160.2 | NP_001191089.1 | 1 - 2 | 123 | 96 |
GenBank mRNA | BC010671.1 | - | 1 - 2 | 123 | 96 |