The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021570.3 | NP_067545.3 | 2 - 3 | 699 | 62 |
GenBank mRNA | AF213356.1 | - | 2 - 3 | 691 | 62 |
BC009458.1 | - | 1 - 2 | 546 | 62 | |
BC064363.1 | - | 2 - 3 | 744 | 62 |