Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRTCAP3
Entrez Gene ID
200634
Gene Name
keratinocyte associated protein 3
Gene Aliases
KCP3
Chromosome Location
Chr.2: 27442366 - 27446481 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168364.1 NP_001161836.1 4 - 5 527 63
NM_001321325.1 NP_001308254.1 4 - 5 527 63
NM_173853.3 NP_776252.2 4 - 5 527 63
GenBank mRNA AY157576.1 - 4 - 5 497 63
AY358993.1 - 3 - 4 585 63
BC101687.1 - 4 - 5 526 63
BC101689.1 - 4 - 5 527 63
BC143574.1 - 4 - 5 526 63
CB160871.1 - 3 - 4 63

View Full Product Details