The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243963.1 | NP_001230892.1 | 3 - 4 | 656 | 164 |
NM_020871.3 | NP_065922.3 | 3 - 4 | 656 | 164 | |
XM_006724724.3 | XP_006724787.2 | 3 - 4 | 659 | 164 | |
XM_017029696.1 | XP_016885185.1 | 3 - 4 | 665 | 164 | |
XM_017029697.1 | XP_016885186.1 | 3 - 4 | 666 | 164 | |
GenBank mRNA | BC125224.1 | - | 3 - 4 | 649 | 164 |
BX647193.1 | - | 3 - 4 | 656 | 164 |