The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098531.2 | NP_001092001.1 | 12 - 13 | 1795 | 64 |
NM_001098532.2 | NP_001092002.1 | 11 - 12 | 1559 | 64 | |
NM_006105.5 | NP_006096.2 | 12 - 13 | 1329 | 64 | |
XM_011537752.1 | XP_011536054.1 | 12 - 13 | 1812 | 64 | |
XM_011537755.1 | XP_011536057.1 | 12 - 13 | 1374 | 64 | |
XM_011537758.2 | XP_011536060.1 | 12 - 13 | 1424 | 64 | |
GenBank mRNA | AF103905.1 | - | 11 - 12 | 1119 | 64 |
AK092448.1 | - | 12 - 13 | 1767 | 64 | |
AK125545.1 | - | 8 - 9 | 812 | 64 | |
AK290230.1 | - | 13 - 14 | 1460 | 64 | |
AK299029.1 | - | 12 - 13 | 1423 | 64 | |
BC017728.1 | - | 12 - 13 | 1409 | 64 | |
BC092404.1 | - | 12 - 13 | 1398 | 64 |