Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACSM3
Entrez Gene ID
6296
Gene Name
acyl-CoA synthetase medium-chain family member 3
Gene Aliases
SA, SAH
Chromosome Location
Chr.16: 20763990 - 20797157 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005622.3 NP_005613.2 11 - 12 1645 84
XM_011545911.1 XP_011544213.1 12 - 13 1750 84
XM_017023523.1 XP_016879012.1 11 - 12 1639 84
XM_017023524.1 XP_016879013.1 10 - 11 1509 84
GenBank mRNA AK223139.1 - 11 - 12 1614 84
AK290322.1 - 11 - 12 1614 84
AK296122.1 - 11 - 12 1444 84
BI826935.1 - 5 - 6 520 84
BU170035.1 - 5 - 6 497 84
D16350.1 - 10 - 11 1485 84

View Full Product Details