Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACSM3
Entrez Gene ID
6296
Gene Name
acyl-CoA synthetase medium-chain family member 3
Gene Aliases
SA, SAH
Chromosome Location
Chr.16: 20763990 - 20797157 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005622.3 NP_005613.2 12 - 13 1738 163
XM_011545911.1 XP_011544213.1 13 - 14 1843 163
XM_017023523.1 XP_016879012.1 12 - 13 1732 163
GenBank mRNA AK223139.1 - 12 - 13 1707 163
AK296122.1 - 12 - 13 1537 163
BI826935.1 - 6 - 7 613 163
BM828310.1 - 2 - 3 133 163
BU170035.1 - 6 - 7 590 163
D16350.1 - 11 - 12 1578 163

View Full Product Details