The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002296.3 | NP_002287.2 | 7 - 8 | 1066 | 110 |
NM_194442.2 | NP_919424.1 | 7 - 8 | 1056 | 110 | |
XM_005273125.2 | XP_005273182.1 | 7 - 8 | 1022 | 110 | |
XM_011544185.2 | XP_011542487.1 | 7 - 8 | 1468 | 110 | |
GenBank mRNA | AB209514.1 | - | 7 - 8 | 973 | 110 |
AK222834.1 | - | 7 - 8 | 953 | 110 | |
AK303589.1 | - | 7 - 8 | 972 | 110 | |
AK312258.1 | - | 7 - 8 | 1007 | 110 | |
BC020079.1 | - | 7 - 8 | 988 | 110 |