Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPNE8
Entrez Gene ID
144402
Gene Name
copine 8
Gene Aliases
-
Chromosome Location
Chr.12: 38652200 - 38908593 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153634.2 NP_705898.1 16 - 17 1348 67
XM_017018851.1 XP_016874340.1 17 - 18 2321 67
XM_017018852.1 XP_016874341.1 16 - 17 1294 67
GenBank mRNA AK074223.1 - 2 - 3 210 67
AK094998.1 - 4 - 5 922 67
AK098593.1 - 16 - 17 1279 67
AK307908.1 - 16 - 17 1319 67
AY177785.1 - 16 - 17 1264 67
BC046366.1 - 4 - 5 665 67
BC110577.1 - 16 - 17 1339 67
BC110578.1 - 16 - 17 1339 67
BX538055.1 - 16 - 17 1362 67

View Full Product Details