The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153634.2 | NP_705898.1 | 19 - 20 | 1586 | 125 |
XM_017018851.1 | XP_016874340.1 | 20 - 21 | 2559 | 125 | |
XM_017018852.1 | XP_016874341.1 | 19 - 20 | 1532 | 125 | |
GenBank mRNA | AK074223.1 | - | 5 - 6 | 448 | 125 |
AK094998.1 | - | 7 - 8 | 1160 | 125 | |
AK098593.1 | - | 19 - 20 | 1517 | 125 | |
AK307908.1 | - | 19 - 20 | 1557 | 125 | |
AY177785.1 | - | 19 - 20 | 1502 | 125 | |
BC046366.1 | - | 7 - 8 | 903 | 125 | |
BC110577.1 | - | 19 - 20 | 1577 | 125 | |
BC110578.1 | - | 19 - 20 | 1577 | 125 | |
BX538055.1 | - | 19 - 20 | 1600 | 125 |