The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153634.2 | NP_705898.1 | 7 - 8 | 557 | 125 |
XM_011537951.2 | XP_011536253.1 | 7 - 8 | 3532 | 125 | |
XM_011537952.2 | XP_011536254.1 | 7 - 8 | 3532 | 125 | |
XM_017018851.1 | XP_016874340.1 | 8 - 9 | 1530 | 125 | |
XM_017018852.1 | XP_016874341.1 | 7 - 8 | 503 | 125 | |
GenBank mRNA | AK098593.1 | - | 7 - 8 | 487 | 125 |
AK307908.1 | - | 7 - 8 | 530 | 125 | |
AY177785.1 | - | 7 - 8 | 473 | 125 | |
BC068040.1 | - | 7 - 8 | 521 | 125 | |
BC110577.1 | - | 7 - 8 | 548 | 125 | |
BC110578.1 | - | 7 - 8 | 548 | 125 | |
BX538055.1 | - | 7 - 8 | 571 | 125 |