The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130782.2 | NP_570138.1 | 2 - 3 | 392 | 151 |
GenBank mRNA | AF076642.1 | - | 2 - 3 | 366 | 151 |
AF268036.1 | - | 2 - 3 | 378 | 151 | |
AK057114.1 | - | 2 - 3 | 385 | 151 | |
AK315377.1 | - | 2 - 3 | 385 | 151 | |
AV715708.1 | - | 2 - 3 | 398 | 151 | |
BC020632.1 | - | 2 - 3 | 384 | 151 |