The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288622.1 | NP_001275551.1 | 8 - 9 | 1230 | 98 |
NM_001288623.1 | NP_001275552.1 | 8 - 9 | 1175 | 98 | |
NM_138468.5 | NP_612477.3 | 9 - 10 | 1357 | 98 | |
GenBank mRNA | AK055978.1 | - | 8 - 9 | 972 | 98 |
AK092310.1 | - | 8 - 9 | 1230 | 98 | |
AL833904.1 | - | 9 - 10 | 1014 | 98 | |
BC063876.1 | - | 8 - 9 | 888 | 98 |