The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001194999.1 | NP_001181928.1 | 6 - 7 | 1006 | 78 |
NM_001195000.1 | NP_001181929.1 | 5 - 6 | 865 | 78 | |
NM_001310213.1 | NP_001297142.1 | 6 - 7 | 981 | 78 | |
NM_001531.2 | NP_001522.1 | 6 - 7 | 1141 | 78 | |
XM_006711286.3 | XP_006711349.1 | 6 - 7 | 1100 | 78 | |
XM_011509463.2 | XP_011507765.1 | 5 - 6 | 913 | 78 | |
GenBank mRNA | AF010446.1 | - | 4 - 5 | 709 | 78 |
AF031469.1 | - | 4 - 5 | 692 | 78 | |
AJ249778.1 | - | 5 - 6 | 985 | 78 | |
AK222910.1 | - | 5 - 6 | 994 | 78 | |
AK290374.1 | - | 4 - 5 | 729 | 78 | |
BC012485.1 | - | 5 - 6 | 850 | 78 | |
BX415555.2 | - | 5 - 6 | 817 | 78 | |
U22963.1 | - | 5 - 6 | 985 | 78 |