The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007365.2 | NP_031391.2 | 13 - 14 | 1637 | 68 |
XM_017000148.1 | XP_016855637.1 | 5 - 6 | 811 | 68 | |
GenBank mRNA | AB023211.1 | - | 13 - 14 | 1619 | 68 |
AB030176.1 | - | 13 - 14 | 1619 | 68 | |
AK123164.1 | - | 1 - 2 | 502 | 68 | |
AK127006.1 | - | 4 - 5 | 1911 | 68 | |
AK295783.1 | - | 10 - 11 | 1289 | 68 |