The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281302.1 | NP_001268231.1 | 5 - 6 | 779 | 124 |
NM_032380.4 | NP_115756.2 | 4 - 5 | 661 | 124 | |
NM_170681.2 | NP_733781.1 | 4 - 5 | 661 | 124 | |
NM_170691.2 | NP_733792.1 | 4 - 5 | 661 | 124 | |
NR_104006.1 | 4 - 5 | 661 | 124 | ||
XM_011543691.2 | XP_011541993.1 | 4 - 5 | 444 | 124 | |
XM_017009986.1 | XP_016865475.1 | 4 - 5 | 555 | 124 | |
GenBank mRNA | AF111808.1 | - | 4 - 5 | 411 | 124 |
AF367997.1 | - | 4 - 5 | 507 | 124 | |
AK025314.1 | - | 4 - 5 | 438 | 124 | |
AK307839.1 | - | 4 - 5 | 351 | 124 | |
AK308422.1 | - | 4 - 5 | 124 | ||
AK315705.1 | - | 4 - 5 | 440 | 124 | |
AY453587.1 | - | 4 - 5 | 405 | 124 | |
BC015712.1 | - | 5 - 6 | 513 | 124 | |
BC030612.1 | - | 4 - 5 | 410 | 124 |