Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC17
Entrez Gene ID
55761
Gene Name
tetratricopeptide repeat domain 17
Gene Aliases
-
Chromosome Location
Chr.11: 43358885 - 43494933 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307943.1 NP_001294872.1 10 - 11 1402 94
NM_018259.5 NP_060729.2 10 - 11 1402 94
XM_005253016.1 XP_005253073.1 10 - 11 1402 94
XM_006718263.1 XP_006718326.1 8 - 9 1085 94
XM_011520218.2 XP_011518520.1 10 - 11 1402 94
XM_017018010.1 XP_016873499.1 10 - 11 1402 94
GenBank mRNA AK001540.1 - 5 - 6 637 94
AK001752.1 - 10 - 11 1355 94
BC017238.2 - 10 - 11 1351 94
BC033000.1 - 10 - 11 1348 94
BX640972.1 - 5 - 6 1242 94

View Full Product Details