Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TTC17
Entrez Gene ID
55761
Gene Name
tetratricopeptide repeat domain 17
Gene Aliases
-
Chromosome Location
Chr.11: 43358885 - 43494933 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307943.1 NP_001294872.1 14 - 15 1913 152
NM_018259.5 NP_060729.2 14 - 15 1913 152
XM_005253016.1 XP_005253073.1 14 - 15 1913 152
XM_006718263.1 XP_006718326.1 12 - 13 1596 152
XM_011520218.2 XP_011518520.1 14 - 15 1913 152
XM_017018010.1 XP_016873499.1 14 - 15 1913 152
GenBank mRNA AK001540.1 - 9 - 10 1148 152
AK001752.1 - 14 - 15 1866 152
AK074253.1 - 4 - 5 430 152
BC017238.2 - 14 - 15 1862 152
BC033000.1 - 14 - 15 1859 152
BX640972.1 - 9 - 10 1753 152

View Full Product Details