The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018259.5 | NP_060729.2 | 19 - 20 | 2860 | 81 |
XM_005253016.1 | XP_005253073.1 | 20 - 21 | 3031 | 81 | |
XM_006718263.1 | XP_006718326.1 | 18 - 19 | 2714 | 81 | |
XM_011520218.2 | XP_011518520.1 | 20 - 21 | 3031 | 81 | |
GenBank mRNA | AK023161.1 | - | 4 - 5 | 660 | 81 |
AK074253.1 | - | 9 - 10 | 1377 | 81 | |
BC017238.2 | - | 19 - 20 | 2809 | 81 |