The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001307943.1 | NP_001294872.1 | 6 - 7 | 849 | 56 |
NM_018259.5 | NP_060729.2 | 6 - 7 | 849 | 56 | |
XM_005253016.1 | XP_005253073.1 | 6 - 7 | 849 | 56 | |
XM_006718263.1 | XP_006718326.1 | 4 - 5 | 532 | 56 | |
XM_011520218.2 | XP_011518520.1 | 6 - 7 | 849 | 56 | |
XM_017018010.1 | XP_016873499.1 | 6 - 7 | 849 | 56 | |
GenBank mRNA | AK001752.1 | - | 6 - 7 | 802 | 56 |
BC017238.2 | - | 6 - 7 | 798 | 56 | |
BC033000.1 | - | 6 - 7 | 795 | 56 | |
BX640972.1 | - | 1 - 2 | 689 | 56 |