Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYTH4
Entrez Gene ID
27128
Gene Name
cytohesin 4
Gene Aliases
CYT4, DJ63G5.1, PSCD4
Chromosome Location
Chr.22: 37282383 - 37315348 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011530148.1 XP_011528450.1 8 - 9 735 89
GenBank mRNA AK024428.1 - 8 - 9 740 89

View Full Product Details