The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001313943.1 | NP_001300872.1 | 1 - 2 | 278 | 68 |
NM_001313944.1 | NP_001300873.1 | 1 - 2 | 278 | 68 | |
NM_001313946.1 | NP_001300875.1 | 1 - 2 | 278 | 68 | |
NM_001313947.1 | NP_001300876.1 | 1 - 2 | 278 | 68 | |
NM_001664.3 | NP_001655.1 | 1 - 2 | 278 | 68 | |
GenBank mRNA | AK130066.1 | - | 1 - 2 | 274 | 68 |
AK130808.1 | - | 1 - 2 | 154 | 68 | |
AK222556.1 | - | 1 - 2 | 170 | 68 | |
AK296654.1 | - | 1 - 2 | 154 | 68 | |
AW403464.1 | - | 1 - 2 | 155 | 68 | |
BC001360.2 | - | 1 - 2 | 110 | 68 | |
BC005976.1 | - | 1 - 2 | 115 | 68 | |
BQ721697.1 | - | 1 - 2 | 151 | 68 | |
BU509012.1 | - | 1 - 2 | 109 | 68 | |
BX647063.1 | - | 1 - 2 | 174 | 68 | |
L25080.1 | - | 1 - 2 | 149 | 68 | |
X05026.1 | - | 1 - 2 | 156 | 68 |