The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018945.3 | NP_061818.1 | 10 - 11 | 1250 | 60 |
XM_005266931.3 | XP_005266988.1 | 9 - 10 | 4099 | 60 | |
XM_011535755.2 | XP_011534057.1 | 11 - 12 | 1079 | 60 | |
XM_017010741.1 | XP_016866230.1 | 9 - 10 | 1885 | 60 | |
GenBank mRNA | AB038040.1 | - | 10 - 11 | 1252 | 60 |
AB209990.1 | - | 9 - 10 | 1356 | 60 | |
AJ251860.1 | - | 10 - 11 | 1244 | 60 | |
AK290582.1 | - | 10 - 11 | 1283 | 60 | |
BC075082.2 | - | 10 - 11 | 973 | 60 | |
BC075083.2 | - | 10 - 11 | 977 | 60 | |
EF108327.1 | - | 9 - 10 | 1358 | 60 |