The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018945.3 | NP_061818.1 | 2 - 3 | 384 | 61 |
XM_011535755.2 | XP_011534057.1 | 3 - 4 | 213 | 61 | |
GenBank mRNA | AB038040.1 | - | 2 - 3 | 386 | 61 |
AJ251860.1 | - | 2 - 3 | 378 | 61 | |
AK290582.1 | - | 2 - 3 | 417 | 61 | |
BC070267.1 | - | 2 - 3 | 316 | 61 | |
BC075082.2 | - | 2 - 3 | 107 | 61 | |
BC075083.2 | - | 2 - 3 | 111 | 61 |