The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018945.3 | NP_061818.1 | 3 - 4 | 463 | 65 |
XM_005266931.3 | XP_005266988.1 | 2 - 3 | 3312 | 65 | |
XM_011535755.2 | XP_011534057.1 | 4 - 5 | 292 | 65 | |
XM_017010741.1 | XP_016866230.1 | 2 - 3 | 1098 | 65 | |
GenBank mRNA | AB038040.1 | - | 3 - 4 | 465 | 65 |
AB209990.1 | - | 2 - 3 | 569 | 65 | |
AJ251860.1 | - | 3 - 4 | 457 | 65 | |
AK290582.1 | - | 3 - 4 | 496 | 65 | |
BC075082.2 | - | 3 - 4 | 186 | 65 | |
BC075083.2 | - | 3 - 4 | 190 | 65 | |
EF108327.1 | - | 2 - 3 | 571 | 65 |