The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318024.1 | NP_001304953.1 | 10 - 11 | 1087 | 105 |
NM_013385.4 | NP_037517.1 | 10 - 11 | 1068 | 105 | |
XM_011530147.2 | XP_011528449.1 | 10 - 11 | 955 | 105 | |
XM_011530148.1 | XP_011528450.1 | 11 - 12 | 1032 | 105 | |
GenBank mRNA | AF075458.1 | - | 10 - 11 | 997 | 105 |
AF125349.1 | - | 10 - 11 | 1068 | 105 | |
AK304453.1 | - | 10 - 11 | 988 | 105 | |
AK315052.1 | - | 10 - 11 | 904 | 105 | |
BC041161.1 | - | 10 - 11 | 926 | 105 | |
CR456551.1 | - | 10 - 11 | 917 | 105 |