The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000747.2 | NP_000738.2 | 3 - 4 | 286 | 63 |
GenBank mRNA | AK298938.1 | - | 2 - 3 | 140 | 63 |
BC011371.1 | - | 3 - 4 | 274 | 63 | |
BC023553.2 | - | 3 - 4 | 286 | 63 | |
X14830.1 | - | 3 - 4 | 261 | 63 |