Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HPN
Entrez Gene ID
3249
Gene Name
hepsin
Gene Aliases
TMPRSS1
Chromosome Location
Chr.19: 35040506 - 35066573 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002151.2 NP_002142.1 13 - 14 1457 74
NM_182983.2 NP_892028.1 12 - 13 2037 74
XM_005258838.4 XP_005258895.2 12 - 13 1535 74
XM_006723181.3 XP_006723244.2 12 - 13 1411 74
XM_017026731.1 XP_016882220.1 12 - 13 1710 74
GenBank mRNA BC025716.1 - 12 - 13 1409 74
M18930.1 - 13 - 14 1457 74
X07002.1 - 7 - 8 873 74
X07732.1 - 12 - 13 2037 74

View Full Product Details