Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HPN
Entrez Gene ID
3249
Gene Name
hepsin
Gene Aliases
TMPRSS1
Chromosome Location
Chr.19: 35040506 - 35066573 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002151.2 NP_002142.1 5 - 6 401 114
NM_182983.2 NP_892028.1 4 - 5 981 114
XM_005258838.4 XP_005258895.2 4 - 5 479 114
XM_006723181.3 XP_006723244.2 4 - 5 355 114
XM_017026731.1 XP_016882220.1 4 - 5 654 114
GenBank mRNA AK315655.1 - 4 - 5 334 114
BC025716.1 - 4 - 5 353 114
EU644753.1 - 3 - 4 156 114
M18930.1 - 5 - 6 401 114
X07732.1 - 4 - 5 981 114

View Full Product Details