The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015993.2 | NP_057077.1 | 1 - 2 | 268 | 114 |
GenBank mRNA | AF137386.1 | - | 1 - 2 | 264 | 114 |
AK130584.1 | - | 1 - 2 | 260 | 114 | |
AK289599.1 | - | 1 - 2 | 245 | 114 | |
AK313910.1 | - | 1 - 2 | 268 | 114 | |
BC002760.2 | - | 1 - 2 | 231 | 114 | |
DA366260.1 | - | 1 - 2 | 269 | 114 |