The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014652.3 | NP_055467.3 | 6 - 7 | 2187 | 76 |
GenBank mRNA | AB018267.1 | - | 6 - 7 | 2054 | 76 |
AF267987.1 | - | 6 - 7 | 1392 | 76 | |
AK290173.1 | - | 6 - 7 | 1815 | 76 | |
AL117501.1 | - | 5 - 6 | 1184 | 76 | |
BC008194.1 | - | 6 - 7 | 2022 | 76 |