The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146094.1 | NP_001139566.1 | 1 - 2 | 96 | |
NM_001146096.1 | NP_001139568.1 | 1 - 2 | 158 | 109 | |
NM_014967.4 | NP_055782.3 | 1 - 2 | 96 | ||
XM_005254232.4 | XP_005254289.1 | 1 - 2 | 153 | 109 | |
XM_011521370.2 | XP_011519672.1 | 1 - 2 | 145 | 109 | |
XM_011521371.1 | XP_011519673.1 | 1 - 2 | 130 | 109 | |
XM_011521372.2 | XP_011519674.1 | 1 - 2 | 96 | ||
XM_017022012.1 | XP_016877501.1 | 1 - 2 | 96 | ||
XM_017022013.1 | XP_016877502.1 | 1 - 2 | 130 | 109 | |
GenBank mRNA | AB023235.2 | - | 1 - 2 | 96 | |
AK290987.1 | - | 1 - 2 | 96 | ||
BP310263.1 | - | 1 - 2 | 116 | 109 | |
CR936727.1 | - | 1 - 2 | 125 | 109 | |
CR936734.1 | - | 1 - 2 | 96 | ||
DA841207.1 | - | 1 - 2 | 96 |