The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016215.4 | NP_057299.1 | 7 - 8 | 939 | 135 |
NM_201446.2 | NP_958854.1 | 5 - 6 | 493 | 135 | |
NR_045110.1 | 6 - 7 | 720 | 135 | ||
NR_045111.1 | 6 - 7 | 723 | 135 | ||
NR_046367.1 | 6 - 7 | 1316 | 135 | ||
XM_011518764.1 | XP_011517066.1 | 7 - 8 | 1640 | 135 | |
XM_011518765.1 | XP_011517067.1 | 7 - 8 | 1637 | 135 | |
XM_011518766.1 | XP_011517068.1 | 8 - 9 | 1856 | 135 | |
GenBank mRNA | AB125649.1 | - | 5 - 6 | 475 | 135 |
AF186111.1 | - | 4 - 5 | 424 | 135 | |
AK091964.1 | - | 6 - 7 | 1316 | 135 | |
AL512735.1 | - | 6 - 7 | 720 | 135 | |
AY358902.1 | - | 7 - 8 | 892 | 135 | |
AY358903.1 | - | 6 - 7 | 730 | 135 | |
BC012377.1 | - | 5 - 6 | 473 | 135 | |
BC088371.1 | - | 6 - 7 | 713 | 135 | |
BM919940.1 | - | 5 - 6 | 493 | 135 | |
KC485578.1 | - | 4 - 5 | 405 | 135 |