Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIF19
Entrez Gene ID
124602
Gene Name
kinesin family member 19
Gene Aliases
KIF19A
Chromosome Location
Chr.17: 74326212 - 74355820 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153209.3 NP_694941.2 3 - 4 366 57
XM_005257029.1 XP_005257086.1 3 - 4 366 57
XM_005257030.1 XP_005257087.1 3 - 4 366 57
XM_017024150.1 XP_016879639.1 3 - 4 366 57
XM_017024151.1 XP_016879640.1 3 - 4 366 57
XM_017024152.1 XP_016879641.1 3 - 4 366 57
XM_017024153.1 XP_016879642.1 3 - 4 366 57
XM_017024154.1 XP_016879643.1 3 - 4 366 57
XM_017024155.1 XP_016879644.1 3 - 4 366 57
XM_017024156.1 XP_016879645.1 3 - 4 366 57
XM_017024157.1 XP_016879646.1 3 - 4 366 57
XM_017024158.1 XP_016879647.1 3 - 4 366 57
GenBank mRNA AK094619.1 - 3 - 4 321 57
BC026362.1 - 3 - 4 347 57
BC093631.1 - 3 - 4 252 57
BC104852.1 - 3 - 4 252 57
BC110989.1 - 3 - 4 330 57
BC143327.1 - 3 - 4 253 57
DA955730.1 - 3 - 4 366 57

View Full Product Details