Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SAMD4B
Entrez Gene ID
55095
Gene Name
sterile alpha motif domain containing 4B
Gene Aliases
SMGB, Smaug2
Chromosome Location
Chr.19: 39342459 - 39390748 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303614.1 NP_001290543.1 11 - 12 2297 78
NM_018028.3 NP_060498.2 14 - 15 3018 78
XM_011527057.2 XP_011525359.1 12 - 13 2308 78
XM_011527060.2 XP_011525362.1 12 - 13 3410 78
XM_011527061.2 XP_011525363.1 11 - 12 2222 78
XM_011527062.2 XP_011525364.1 11 - 12 2283 78
XM_011527063.2 XP_011525365.1 12 - 13 2307 78
XM_011527064.2 XP_011525366.1 12 - 13 2308 78
XM_017026918.1 XP_016882407.1 11 - 12 2242 78
XM_017026919.1 XP_016882408.1 13 - 14 2398 78
XM_017026920.1 XP_016882409.1 13 - 14 2420 78
XM_017026921.1 XP_016882410.1 11 - 12 2283 78
XM_017026922.1 XP_016882411.1 13 - 14 2400 78
XM_017026923.1 XP_016882412.1 13 - 14 7938 78
GenBank mRNA AK091041.1 - 1 - 2 95 78
BC039652.1 - 7 - 8 995 78
BC054518.1 - 14 - 15 3009 78
BC065211.1 - 8 - 9 1305 78
EF601121.1 - 14 - 15 3005 78

View Full Product Details