The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199898.1 | NP_001186827.1 | 6 - 7 | 811 | 67 |
NM_001199899.1 | NP_001186828.1 | 5 - 6 | 610 | 67 | |
NM_002611.4 | NP_002602.2 | 5 - 6 | 773 | 67 | |
GenBank mRNA | AK055119.1 | - | 6 - 7 | 811 | 67 |
AK290522.1 | - | 5 - 6 | 672 | 67 | |
AK293600.1 | - | 5 - 6 | 587 | 67 | |
AK297089.1 | - | 5 - 6 | 560 | 67 | |
BC005811.2 | - | 5 - 6 | 661 | 67 | |
BC040478.1 | - | 5 - 6 | 716 | 67 | |
BC063137.1 | - | 5 - 6 | 687 | 67 | |
L42451.1 | - | 5 - 6 | 660 | 67 |