The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040455.1 | NP_001035545.1 | 13 - 14 | 1316 | 64 |
GenBank mRNA | AF151799.1 | - | 14 - 15 | 1421 | 64 |
AK075137.1 | - | 4 - 5 | 294 | 64 | |
AK292501.1 | - | 13 - 14 | 1796 | 64 | |
AK292974.1 | - | 13 - 14 | 1307 | 64 | |
AK301501.1 | - | 5 - 6 | 586 | 64 | |
AK303023.1 | - | 13 - 14 | 1272 | 64 | |
AY358442.1 | - | 13 - 14 | 1325 | 64 | |
BC114522.1 | - | 14 - 15 | 1336 | 64 | |
BX648437.1 | - | 5 - 6 | 414 | 64 |