Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SIDT2
Entrez Gene ID
51092
Gene Name
SID1 transmembrane family member 2
Gene Aliases
CGI-40
Chromosome Location
Chr.11: 117179223 - 117197445 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040455.1 NP_001035545.1 17 - 18 1656 67
GenBank mRNA AF151799.1 - 18 - 19 1761 67
AK075137.1 - 8 - 9 634 67
AK292501.1 - 17 - 18 2136 67
AK292974.1 - 17 - 18 1647 67
AK301501.1 - 8 - 9 852 67
AK303023.1 - 17 - 18 1612 67
AY358442.1 - 17 - 18 1665 67
BC114522.1 - 18 - 19 1676 67
KC109742.1 - 17 - 18 1612 67

View Full Product Details