The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040455.1 | NP_001035545.1 | 21 - 22 | 2097 | 74 |
GenBank mRNA | AF151799.1 | - | 22 - 23 | 2202 | 74 |
AK075137.1 | - | 12 - 13 | 1075 | 74 | |
AK075471.1 | - | 3 - 4 | 289 | 74 | |
AK292501.1 | - | 21 - 22 | 2577 | 74 | |
AK303023.1 | - | 21 - 22 | 2053 | 74 | |
AY358442.1 | - | 21 - 22 | 2106 | 74 | |
BC114522.1 | - | 22 - 23 | 2117 | 74 | |
BX648437.1 | - | 13 - 14 | 1195 | 74 |