The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040455.1 | NP_001035545.1 | 6 - 7 | 744 | 62 |
GenBank mRNA | AF151799.1 | - | 6 - 7 | 786 | 62 |
AK292501.1 | - | 6 - 7 | 1224 | 62 | |
AK292974.1 | - | 6 - 7 | 735 | 62 | |
AK303023.1 | - | 6 - 7 | 700 | 62 | |
AY358442.1 | - | 6 - 7 | 753 | 62 | |
BC114522.1 | - | 6 - 7 | 773 | 62 | |
KC109742.1 | - | 6 - 7 | 700 | 62 |