The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040455.1 | NP_001035545.1 | 9 - 10 | 999 | 101 |
GenBank mRNA | AF151799.1 | - | 9 - 10 | 1041 | 101 |
AK292501.1 | - | 9 - 10 | 1479 | 101 | |
AK292974.1 | - | 9 - 10 | 990 | 101 | |
AK301501.1 | - | 1 - 2 | 269 | 101 | |
AK303023.1 | - | 9 - 10 | 955 | 101 | |
AY358442.1 | - | 9 - 10 | 1008 | 101 | |
BC114522.1 | - | 9 - 10 | 1028 | 101 | |
BX648437.1 | - | 1 - 2 | 97 | 101 | |
KC109742.1 | - | 9 - 10 | 955 | 101 |