The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004334.2 | NP_004325.2 | 6 - 7 | 899 | 86 |
XM_005248185.1 | XP_005248242.1 | 6 - 7 | 899 | 86 | |
XM_005248186.1 | XP_005248243.1 | 6 - 7 | 899 | 86 | |
XM_011513878.2 | XP_011512180.1 | 6 - 7 | 899 | 86 | |
XM_011513879.1 | XP_011512181.1 | 6 - 7 | 899 | 86 | |
XM_011513881.2 | XP_011512183.1 | 6 - 7 | 1809 | 86 | |
XM_017008565.1 | XP_016864054.1 | 6 - 7 | 899 | 86 | |
XM_017008566.1 | XP_016864055.1 | 6 - 7 | 899 | 86 | |
XM_017008567.1 | XP_016864056.1 | 6 - 7 | 1318 | 86 | |
GenBank mRNA | AB175627.1 | - | 6 - 7 | 530 | 86 |
AK312497.1 | - | 6 - 7 | 776 | 86 | |
BC012095.1 | - | 6 - 7 | 774 | 86 | |
BT019502.1 | - | 6 - 7 | 704 | 86 | |
BU732655.1 | - | 3 - 4 | 86 | ||
D21878.1 | - | 6 - 7 | 831 | 86 |