The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001185072.2 | NP_001172001.1 | 4 - 4 | 1472 | 96 |
NM_001185073.2 | NP_001172002.1 | 3 - 3 | 1433 | 96 | |
NM_012129.4 | NP_036261.1 | 3 - 3 | 1429 | 96 | |
GenBank mRNA | AL136770.1 | - | 3 - 3 | 1281 | 96 |
BC036754.2 | - | 2 - 2 | 1218 | 96 | |
BC068532.1 | - | 3 - 3 | 1315 | 96 |